missing translation for 'onlineSavingsMsg'
Learn More

Sodium chromate tetrahydrate, 99+%

Product Code. 11498107
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498107 500 g 500g
11488107 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11498107 Supplier Thermo Scientific Alfa Aesar Supplier No. A17499.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10034-82-9 ∣ CrH8Na2O8 ∣ 234.032 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications

Sodium chromate tetrahydrate is used as a corrosion inhibitor, wood preservative and an auxiliary in textile industry. It serves as an oxidant, which is used to prepare carboxylic acids and ketones from primary and secondary alcohols respectively. Further, it is used as a starting material to prepare other chromium compounds and as a catalyst in organic synthesis especially pharmaceuticals and agro chemicals.

Solubility

Soluble in water.

Notes

Hygroscopic. Incompatible withstrong reducing agents, organic materials, strong acids, combustible materials and powdered metals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-82-9
Molecular Formula CrH8Na2O8
Molecular Weight (g/mol) 234.032
MDL Number MFCD00149167
InChI Key AVHUVTPOXZEIRG-UHFFFAOYSA-N
Synonym sodium chromate tetrahydrate, cro4.2na.4h2o, acmc-20ajol, disodium chromate tetrahydrate, ksc492q3d, disodium chromate 2-tetrahydrate, sodium chromate tetrahydrate, reagent grade, sodium chromate tetrahydrate, p.a
PubChem CID 22146515
IUPAC Name disodium;dioxido(dioxo)chromium;tetrahydrate
SMILES O.O.O.O.[O-][Cr](=O)(=O)[O-].[Na+].[Na+]

Specifications

Physical Form Crystalline Powder and/or Crystals
Quantity 500 g
UN Number UN3288
Merck Index 14,8601
Solubility Information Soluble in water.
Formula Weight 234.04 (161.97 Anhydrous)
Percent Purity ≥99%
Grade Ungraded
Odor Odorless
Density 2.73
Chemical Name or Material Sodium chromate tetrahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.