missing translation for 'onlineSavingsMsg'
Learn More

Sodium Citrate, Dihydrate, Granular, U.S.P. - F.C.C., J.T. Baker™

Product Code. 15568154 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Wide Mouth Amber Glass Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15568154 500 g Wide Mouth Amber Glass Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15568154 Supplier Avantor J.T.Baker Supplier No. 364901
  • ONLINE EXCLUSIVE PRICE
    129.71 €
    107.49 € / 500g
    Save 22.22 €
    17% Off
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 4-3-6132
Molecular Formula C6H9Na3O9
Molecular Weight (g/mol) 294.10
InChI Key NLJMYIDDQXHKNR-UHFFFAOYSA-K
Synonym trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
PubChem CID 71474
ChEBI CHEBI:32142
IUPAC Name trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]

Specifications

Physical Form Granules
Quantity 500 g
Formula Weight 294.1
Percent Purity 99.0% to 100.5%
Grade FCC/USP
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 1.70kg
Chemical Name or Material Sodium Citrate, Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.