missing translation for 'onlineSavingsMsg'
Learn More

Sodium citrate dihydrate, granular, USP/FCC/EP/BP/JP, Fisher Chemical™

Product Code. 15533390 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
12 kg
Unit Size:
12kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15533390 12 kg 12kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15533390 Supplier Thermo Scientific Acros Supplier No. USS47012
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

pH 7.0 to 9.0 (5% aq. soln.)
CAS 4-3-6132
Melting Point 300°C
Boiling Point >300°C
Color White
Appearance Pass Test (EP/BP/JP)
Density 1.67 g/mL
Quantity 12 kg
Loss on Drying 10.0 to 13.0% (JP)
Packaging Plastic
Physical Form Solid
Molecular Formula C6H9Na3O9
Linear Formula NaO2CCH2C(OH)(CO2Na)CH2CO2Na·2H2O
MDL Number MFCD00150031
Merck Index 15, 8737
Solubility Information Soluble in water
InChI Key NLJMYIDDQXHKNR-UHFFFAOYSA-K
SMILES O.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
IUPAC Name trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
Molecular Weight (g/mol) 294.10
Formula Weight 294.1
Grade BP/EP/FCC/JP/USP
Chemical Name or Material Sodium citrate dihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.