missing translation for 'onlineSavingsMsg'
Learn More

Sodium Citrate, TRC

Product Code. 30134117
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134117 500 g 500g
30006830 10 g 10g
30005014 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134117 Supplier Toronto Research Chemicals Supplier No. TRCS655008500G
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-04-2
Molecular Formula C6H5Na3O7
Molecular Weight (g/mol) 258.07
Synonym 2-Hydroxy-1,2,3-propanenetricarboxylic Acid Trisodium Salt Dihydrate, 2-Hydroxy-,1,2,3-propanetricarboxylic Acid Trisodium Salt (9CI), Citric Acid, Trisodium Salt (8CI), Sodium Citrate (Na3C6H5O7) (7CI);2-Hydroxy-1,2,3-propanetricarboxylic Acid Trisodium Salt, 331iii, Citnatin, Citra-lock, Citreme, Citrosodina, Citrosodine, Cystemme, Ional, Natrocitral, P3, Sodium citrate Anhydrous, Trisodium Citrate, Unifine P 3, Urisal
IUPAC Name trisodium;2-hydroxypropane-1,2,3-tricarboxylate
SMILES [Na+].[Na+].[Na+].OC(CC(=O)[O-])(CC(=O)[O-])C(=O)[O-]

Specifications

Quantity 500 g
InChI Formula InChI=1S/C6H8O7.3Na/c7-3(8)1-6(13,5(11)12)2-4(9)10;;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;/q;3*+1/p-3
Formula Weight 257.9728
Percent Purity >95%
Chemical Name or Material Sodium Citrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.