missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Sodium Cromoglicate
Description
Sodium Cromoglicate is a promotes transformation ESCs/iPSCs into pancreatic endocrine islet cells. Facilitates differentiation from endocrine precursors. Also inhibits mast cell degranulation and histamine release. Antiasthmatic.
Specifications
Specifications
| Chemical Name or Material | 5,5'-[(2-Hydroxy-1,3-propanediyl)bis(oxy)]bis[4-oxo-4H-1-benzopyran-2-carboxylic acid] disodium salt |
| CAS | 15826-37-6 |
| Assay Percent Range | >98% |
| Quantity | 50 mg |
| Product Line | Tocris |
| Molecular Formula | C23H14Na2O11 |
| Formula Weight | Observed MW: 512.33 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?