Learn More
Sodium Cromoglicate, Tocris Bioscience™
Promotes ESCs/iPSCs differentiation into pancreatic endocrine islet cells
Brand:  Tocris Bioscience 6424/50
Encompass
Product Code. 15963291
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Sodium Cromoglicate is a promotes transformation ESCs/iPSCs into pancreatic endocrine islet cells. Facilitates differentiation from endocrine precursors. Also inhibits mast cell degranulation and histamine release. Antiasthmatic.
Specifications
Specifications
5,5'-[(2-Hydroxy-1,3-propanediyl)bis(oxy)]bis[4-oxo-4H-1-benzopyran-2-carboxylic acid] disodium salt
15826-37-6
>98%