missing translation for 'onlineSavingsMsg'
Learn More

Sodium cyanate, 85%, pure

Product Code. 10275660 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
Plastic bottle
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10275660 250 g Plastic bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10275660 Supplier Thermo Scientific Acros Supplier No. 180562500
  • ONLINE EXCLUSIVE PRICE
    61.80 €
    46.60 € / 250g
    Save 15.20 €
    24% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 917-61-3 | CNNaO | 65.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 550°C
CAS 917-61-3
Color Beige or White to Yellow
Physical Form Crystalline Powder and/or Chunks
Quantity 250 g
Loss on Drying 3 % max. (5 g, 105°C, 2 hrs)
Packaging Plastic bottle
Assay Percent Range 80% min. (Argentometry)
Molecular Formula CNNaO
Linear Formula NaOCN
MDL Number MFCD00011121
Beilstein 03, IV, 82
Merck Index 15, 8740
Synonym sodium cyanate, zassol, cyanic acid, sodium salt, sodiumcyanate, cyansan, weecon, sodium isocyanate, san-cyan, unii-8ufs3jrv8p, naocn
Solubility Information Solubility in water: 110g/L (20 c). Other solubilities: 0.22g/100g alcohol (0 c),insoluble in ether
InChI Key ZVCDLGYNFYZZOK-UHFFFAOYSA-M
SMILES C(#N)[O-].[Na+]
IUPAC Name sodium;cyanate
Molecular Weight (g/mol) 65.01
PubChem CID 517096
ChEBI CHEBI:38906
Formula Weight 65.01
Percent Purity 85%
Grade Pure
Chemical Name or Material Sodium cyanate, 85%
Show More Show Less
compliance-icons
Product Identifier
  • Sodium cyanate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.