missing translation for 'onlineSavingsMsg'
Learn More

Sodium cyanide, 98%, for analysis, Thermo Scientific™

Product Code. 12644987 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12644987 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 12644987 Supplier Thermo Scientific Acros Supplier No. 194661000
  • ONLINE EXCLUSIVE PRICE
    35.95 €
    33.70 € / 100g
    Save 2.25 €
    6% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 1497.0°C
Melting Point 562.0°C
CAS 143-33-9
Color White
Physical Form Granular Crystalline Powder
Quantity 100 g
Assay Percent Range 97% min. (Argentometry)
Molecular Formula CNNa
Linear Formula NaCN
MDL Number MFCD00003523
Beilstein 02,IV,5
Fieser 04,446; 05,506; 06,535; 07,333; 08,453; 11,481; 13,185
Merck Index 15,8741
Synonym cymag, sodium cyanide, kyanid sodny, cyanogran, nacn, cyanasalt h, cyanide salts, cyanide of sodium, sodium cyanide na cn, cyanobrik
Solubility Information Solubility in water: 370g/L (20°C). Other solubilities: sparingly soluble in ethanol
InChI Key MNWBNISUBARLIT-UHFFFAOYSA-N
SMILES [Na+].[C-]#N
IUPAC Name sodium iminomethanide
Molecular Weight (g/mol) 49.01
PubChem CID 8929
Formula Weight 49
Percent Purity 98%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Sodium cyanide, p.a.
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium cyanide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Substances and mixtures corrosive to metals Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H290-May be corrosive to metals.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P390-Absorb spillage to prevent material damage.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.