missing translation for 'onlineSavingsMsg'
Learn More

Sodium Cyanide, Extra Pure, SLR, Fisher Chemical

Product Code. 10478800 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10478800 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10478800 Supplier Fisher Chemical Supplier No. S/3480/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolitik
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Tekniske data

Boiling Point 1497°C
Melting Point 562°C
CAS 143-33-9
Color White
pH 11 to 12
Physical Form Solid
Quantity 500 g
Molecular Formula CNNa
MDL Number MFCD00003523
Synonym sodium cyanide, cyanogran, cymag, cyanide of sodium, sodium cyanide na cn, nacn, cyanobrik, cyanide salts, kyanid sodny, cyanasalt h
InChI Key MNWBNISUBARLIT-UHFFFAOYSA-N
SMILES [Na+].[C-]#N
IUPAC Name sodium iminomethanide
Molecular Weight (g/mol) 49.01
PubChem CID 8929
Formula Weight 49g/mol
Packaging Plastic powder jar
Vapor Pressure 1 hPa 817
Vis mere Vis mindre
compliance-icons compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • Sodium cyanide
Signalord
  • Danger
Farekategori
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 1
  • Specific target organ toxicity Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Faresætning
  • H290-May be corrosive to metals.
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H372-Causes damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.
Sikkerhedssætning
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P390-Absorb spillage to prevent material damage.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.