missing translation for 'onlineSavingsMsg'
Learn More

Sodium Cyanide, TRC

Product Code. 30412632
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30412632 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30412632 Supplier Toronto Research Chemicals Supplier No. TRCS61540050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyanosodium
CAS 143-33-9
Molecular Formula C N Na
Quantity 50 g
Synonym Cyanasalt H, Cyanasalt S, Cymag, NSC 77379, Natrium Cyanide
InChI Formula InChI=1S/CN.Na/c1-2;
SMILES [Na]C#N
IUPAC Name cyanosodium
Molecular Weight (g/mol) 49.007
Formula Weight 48.993
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.