missing translation for 'onlineSavingsMsg'
Learn More

Sodium cyanoborohydride, 1M solution in THF, AcroSeal™

Product Code. 10386694 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
Product Code. Quantity Packaging unitSize
10386694 800 mL AcroSeal™ Glass Bottle 800mL
10113354 100 mL AcroSeal™ Glass Bottle 100mL
2 options
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Product Code. 10386694

Brand: Thermo Scientific Acros 372458000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

CAS: 25895-60-7 | CH3BNNa | 62.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium cyanoborohydride
Name Note 1M solution in THF
CAS 25895-60-7, 109-99-9
Quantity 800 mL
Molecular Formula CH3BNNa
Linear Formula NaBH3CN
MDL Number MFCD00003516
Flash Point −18°C
Merck Index 15, 8742
Synonym sodium cyanoborohydride, sodium cyanotrihydridoborate, unii-c4i8c58p9t, zlchem 216, sodium cyanoboron 1-
Solubility Information Solubility in water: reacts. Other solubilities: soluble in methanol; soluble in thf, 372g/L, at 28°C, 410g/L at 46°C, 422g/L at 62°C, soluble in diglyme 176g/L at 25°C, slightly soluble in ethanol, insoluble in ether, benzene, hexane
InChI Key CVDUGUOQTVTBJH-UHFFFAOYSA-N
SMILES [Na+].[BH3-]C#N
IUPAC Name sodium;cyanoboron(1-)
Molecular Weight (g/mol) 62.84
PubChem CID 20587905
Formula Weight 62.84
Packaging AcroSeal™ Glass Bottle
Appearance Clear colorless solution
Specific Gravity 0.915
Density 0.9150g/mL
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium cyanoborohydride, 1M solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H411-Toxic to aquatic life with long lasting effects.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.