missing translation for 'onlineSavingsMsg'
Learn More

Sodium cyanoborohydride, 95%

Product Code. 11410960
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410960 5 g 5g
15432977 25 g 25g
11400960 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11410960 Supplier Thermo Scientific Alfa Aesar Supplier No. 087839.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective Reducing Agent For Aldehydes, Ketones, Oximes | CAS: 25895-60-7 | CH3BNNa | 62.84 g/mol

Selective reducing agent for aldehydes, ketones, oximes, enamines. Reductive animation of ketones, aldehydes. Reductive alkylation of amines, hydrazines Sodium cyanoborohydride is used as a reagent involved in the reductive amination of aldehydes, ketones, oximes and enamines. It is also used in the reductive alkylation of amines and hydrazines. It acts as a catalyst in the preparation of phenolate-bridged dilanthanum(III) complex. In addition, it is used in the conversion of imines to amines. It plays an important role in the reductive deoxygenation of ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Moisture Sensitive; Highly flammable
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25895-60-7
Molecular Formula CH3BNNa
Molecular Weight (g/mol) 62.84
MDL Number MFCD00003516
InChI Key CVDUGUOQTVTBJH-UHFFFAOYSA-N
Synonym sodium cyanoborohydride, sodium cyanotrihydridoborate, unii-c4i8c58p9t, zlchem 216, sodium cyanoboron 1-
PubChem CID 20587905
SMILES [Na+].[BH3-]C#N

Specifications

Melting Point >300°C (decomposition)
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 95%
Solubility Information Soluble in water,THF,methanolSoluble in water,diglyme,tetrahydrofuran and methanol. Slightly soluble in dimethyl sulfoxide. Insoluble in diethyl ether.
IUPAC Name sodium;cyanoboron(1-)
Formula Weight 62.84
Sensitivity Moisture sensitive
Density 1.20 g/cm3 at 20°C
Chemical Name or Material Sodium cyanoborohydride
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium cyanotrihydridoborate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 1 B
  • Substance which, in contact with water, emits flammable gases Category 2
Hazard Statement
  • H228-Flammable solid.
  • H261-In contact with water releases flammable gases.
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Sodium cyanotrihydridoborate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.