missing translation for 'onlineSavingsMsg'
Learn More

Sodium cyanoborohydride, 95%

Product Code. 10541231 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10541231 10 g Glass bottle 10g
10082110 50 g Glass bottle 50g
10703504 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10541231 Supplier Thermo Scientific Acros Supplier No. 168550100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

CAS: 25895-60-7 | CH3BNNa | 62.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25895-60-7
Molecular Formula CH3BNNa
Molecular Weight (g/mol) 62.84
MDL Number MFCD00003516
InChI Key CVDUGUOQTVTBJH-UHFFFAOYSA-N
Synonym sodium cyanoborohydride, sodium cyanotrihydridoborate, unii-c4i8c58p9t, zlchem 216, sodium cyanoboron 1-
PubChem CID 20587905
SMILES [Na+].[BH3-]C#N

Specifications

Melting Point >242.0°C
Color White
Physical Form Powder
Quantity 10 g
Assay Percent Range 95%
Linear Formula NaBH3CN
Fieser 04,448; 05,607; 06,537; 07,334; 08,454; 09,424; 10,360; 11,481; 12,445; 14,287; 16,305; 17,388
Merck Index 15, 8742
Solubility Information Solubility in water: 2120g/L at 29°C (slow decomposition). Other solubilities: soluble in methanol; soluble in thf, 372g/L, at 28°C, 410g/L at 46°C, 422g/L at 62°C, soluble in diglyme 176g/L at 25°C, slightly soluble in ethanol, insoluble in ether, b
IUPAC Name sodium;cyanoboron(1-)
Formula Weight 62.84
Percent Purity 95%
Packaging Glass bottle
Chemical Name or Material Sodium cyanoborohydride
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium cyanoborohydride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 1 B
  • Substance which, in contact with water, emits flammable gases Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H261-In contact with water releases flammable gases.
  • H228-Flammable solid.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.