missing translation for 'onlineSavingsMsg'
Learn More

Sodium Cyanoborohydride, TRC

Product Code. 30425801
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
Product Code. Quantity unitSize
30425801 50 g 50g
1 options
This item is not returnable. View return policy

Product Code. 30425801

Brand: Toronto Research Chemicals TRCS61550050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Cyanoborohydride
CAS 25895-60-7
Purity Grade Notes HPLC
Molecular Formula CH3BNNa
Quantity 50 g
Synonym Sodium (T-4)-(Cyano-C)trihydroborate(1-), Sodium borocyanohydride, Sodium Cyanotrihydroborate, Sodium Cyanotrihydroborate(1-)
InChI Formula InChI=1S/CH3BN.Na/c2-1-3;/h2H3;/q-1;+1
SMILES [Na+].[BH3-]C#N
IUPAC Name sodium;cyanoboranuide
Molecular Weight (g/mol) 62.84
Formula Weight 63.0256
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.