missing translation for 'onlineSavingsMsg'
Learn More

Sodium cyclamate, 98%

Product Code. 11422958
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11422958 5000 g 5000g
11412958 250 g 250g
11402958 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11422958 Supplier Thermo Scientific Alfa Aesar Supplier No. A18666.0I
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 139-05-9 | C6H12NNaO3S | 201.22 g/mol

Sodium Cyclamate is an artificial sweetener and is 30 times as saccharose's. It is widely used in pickles, seasoning sauce, cakes, biscuits, bread, ice cream, frozen sucker, popsicles, drinks and so on, with a maximum amount of 0.65g/kg. Secondly, it is used in confect, with a maximum amount of 1.0g/kg. Thirdly, it is used in orange peel, preserved plum, dried arbutus and so on, with the largest amount of 8.0g/kg. It is also used in cosmetics and pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium Cyclamate is an artificial sweetener and is 30 times as saccharose′s. It is widely used in pickles, seasoning sauce, cakes, biscuits, bread, ice cream, frozen sucker, popsicles, drinks and so on, with a maximum amount of 0.65g/kg. Secondly, it is used in confect, with a maximum amount of 1.0g/kg. Thirdly, it is used in orange peel, preserved plum, dried arbutus and so on, with the largest amount of 8.0g/kg. It is also used in cosmetics and pharmaceutical industry.

Solubility
Soluble in water (>=10 g/100 mL at 20°C).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139-05-9
Molecular Formula C6H12NNaO3S
Molecular Weight (g/mol) 201.22
MDL Number MFCD00003827
InChI Key UDIPTWFVPPPURJ-UHFFFAOYSA-M
Synonym sodium cyclamate, sodium cyclohexylsulfamate, sodium n-cyclohexylsulfamate, cyclamate sodium, cyclamic acid sodium salt, assugrin, ibiosuc, suessette, suestamin, sugarin
PubChem CID 23665706
ChEBI CHEBI:82431
SMILES [Na+].[O-]S(=O)(=O)NC1CCCCC1

Specifications

Melting Point >300°C
Quantity 5000 g
Beilstein 4166868
Solubility Information Soluble in water (>=10g/100 mL at 20°C).
IUPAC Name sodium;N-cyclohexylsulfamate
Formula Weight 201.22
Percent Purity 98%
Chemical Name or Material Sodium cyclamate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.