missing translation for 'onlineSavingsMsg'
Learn More

Sodium D-Gluconate, TRC

Product Code. 17967194
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17967194 25 g 25g
17948014 5 g 5g
17957194 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17967194 Supplier Toronto Research Chemicals Supplier No. TRCS63400025G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 527-07-1
Molecular Formula C6H11NaO7
Molecular Weight (g/mol) 218.14
Synonym Sodium Gluconate, D-Gluconic acid, sodium salt (1:1) (ACI), D-Gluconic acid, monosodium salt (9CI), Gluconic acid, monosodium salt, D- (8CI), BVD Addicrete, Clewat GL, Disparlight DV, EMF 1240, GL-P, GL-P (gluconate), Glonsen, Glucon SGA 60, Gluconic acid monosodium salt, Gluconic acid sodium salt, Helshas A, Monosodium gluconate, Pasexon 100T, PMP Sodium Gluconate, Resitard P 608A, Sodium D-gluconate, Sodium gluconate, Sunmorl N 60S
IUPAC Name Sodium gluconate
SMILES [Na+].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(=O)[O-]

Specifications

Quantity 25 g
InChI Formula InChI=1S/C6H12O7.Na/c7-1-2(8)3(9)4(10)5(11)6(12)13;/h2-5,7-11H,1H2,(H,12,13);/q;+1/p-1/t2-,3-,4+,5-;/m1./s1
Formula Weight 218.0402
Percent Purity >95%
Chemical Name or Material Sodium gluconate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.