missing translation for 'onlineSavingsMsg'
Learn More

Sodium D-glucuronate monohydrate, 99%

Product Code. 11348865
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11348865 25 g 25g
11338865 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348865 Supplier Thermo Scientific Alfa Aesar Supplier No. L13985.14
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 207300-70-7 ∣ C6H11NaO8 ∣ 234.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207300-70-7
Molecular Formula C6H11NaO8
Molecular Weight (g/mol) 234.14
MDL Number MFCD00151051
InChI Key ICQRTPLATMRWJI-UHFFFAOYNA-M
Synonym d-glucuronic acid sodium salt hydrate, glv8d2bupd, unii-glv8d2bupd, sodium d-glucuronate, sodium d-glucuronate monohydrate, sodium glucuronate hydrate jan, sodium 2s,3s,4s,5r-2,3,4,5-tetrahydroxy-6-oxohexanoate hydrate, sodium-d-glucuronate, d-glucuronic acid sodium salt
PubChem CID 24892836
SMILES O.[Na+].OC(C=O)C(O)C(O)C(O)C([O-])=O

Specifications

Melting Point 134°C to 138°C
Physical Form Crystalline Powder and/or Crystals
Quantity 25 g
Beilstein 4218577
IUPAC Name sodium;(2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoate;hydrate
Formula Weight 234.14 (216.14 Anhydrous)
Percent Purity 99%
Grade Ungraded
Chemical Name or Material Sodium D-glucuronate monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.