missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Sodium Deoxycholate Monohydrate, 25 g

Product Code. 17851391 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17851391 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17851391 Supplier MP Biomedicals Supplier No. 0219993325
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sodium Deoxycholate Monohydrate is a powerful solubilization agent; used in the solubilization of estrogen synthetase from human placental microsomes.

  • Sodium deoxycholate monohydrate has been used in a study to:
    • Assess a method for the immobilization of lipoxygenase in an alginate-silicate gel matrix
    • Investigate the in vitro dissolution of gall stones

Specifications

Color White
CAS 145224-92-6
Boiling Point Undetermined
Melting Point >300°C
Quantity 25 g
MDL Number MFCD00150750
Beilstein 3643164
Molecular Weight (g/mol) 432.577 g/mol
Percent Purity ≥98%
compliance-icons
Product Identifier
  • Sodium deoxycholate monohydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Sodium deoxycholatemonohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.