missing translation for 'onlineSavingsMsg'
Learn More

Sodium deuteroxide, 40% w/w solution in D{2}O, 99.5%(Isotopic)

Product Code. 11304448
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11304448 50 g 50g
11314448 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11304448 Supplier Thermo Scientific Alfa Aesar Supplier No. 042346.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 14014-06-3 | HNaO | 41.003 g/mol

Sodium deuteroxide is a reagent used in the synthesis of rac Zearalenone-d6. is used in many chemical reactions and industrial practices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14014-06-3
Molecular Formula HNaO
Molecular Weight (g/mol) 41.003
MDL Number MFCD00037669
InChI Key HEMHJVSKTPXQMS-DYCDLGHISA-M
Synonym sodium deuteroxide, sodium hydroxide na od, sodium deuteroxide, 40 wt% in deuterium oxide, naod, sodium hydroxide-d, sodium deuteroxide solution, sodium deuteroxide, 1 m in deuterium oxide, sodium deuteroxide, 0.1 m in deuterium oxide, sodium deuteroxide, 30 wt% in deuterium oxide, sodium hydroxide na od 6ci,7ci,8ci,9ci
PubChem CID 23676750
SMILES [OH-].[Na+]

Specifications

Color Colorless
Physical Form Liquid
Quantity 50 g
Assay Percent Range 99.5%
Solubility Information Fully miscible in water.
Formula Weight 41.01
Odor Odorless
Refractive Index 1.418
Sensitivity Air and moisture sensitive
Appearance Colorless liquid
Density 1.462 g/cm3 at 20°C
Chemical Name or Material Sodium deuteroxide, Isotopic
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.