missing translation for 'onlineSavingsMsg'
Learn More

Sodium deuteroxide, for NMR, 40 wt% solution in D2O, 99+ atom % D

Product Code. 10573694 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Plastic Bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10573694 10 g Plastic Bottle 10g
10377830 50 g Plastic Bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10573694 Supplier Thermo Scientific Acros Supplier No. 175470100
  • ONLINE EXCLUSIVE PRICE
    41.35 €
    40.10 € / 10g
    Save 1.25 €
    3% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 14014-06-3 | HNaO | 41.003 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14014-06-3 , 7789-20-0
Molecular Formula HNaO
Molecular Weight (g/mol) 41.003
MDL Number MFCD00037669
InChI Key HEMHJVSKTPXQMS-DYCDLGHISA-M
Synonym sodium deuteroxide, sodium hydroxide na od, sodium deuteroxide, 40 wt% in deuterium oxide, naod, sodium hydroxide-d, sodium deuteroxide solution, sodium deuteroxide, 1 m in deuterium oxide, sodium deuteroxide, 0.1 m in deuterium oxide, sodium deuteroxide, 30 wt% in deuterium oxide, sodium hydroxide na od 6ci,7ci,8ci,9ci
PubChem CID 23676750
SMILES [OH-].[Na+]

Specifications

Physical Form Liquid
Color Colorless
Density 1.4600g/mL
Deuterium (D) 99.0% min.
Packaging Plastic Bottle
Refractive Index 1.4039
Quantity 10 g
Linear Formula NaOD
Formula Weight 41
Grade NMR
Chemical Name or Material Sodium deuteroxide, 99+ atom % D
Show More Show Less
compliance-icons
Product Identifier
  • Sodium deuteroxide, 30 wt% solution in D2O
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.