missing translation for 'onlineSavingsMsg'
Learn More

Sodium diatrizoate, 99%

Product Code. 15658775 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15658775

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 737-31-5 | C11H8I3N2NaO4 | 635.9 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 737-31-5
Molecular Formula C11H8I3N2NaO4
Molecular Weight (g/mol) 635.9
InChI Key ZEYOIOAKZLALAP-UHFFFAOYSA-M
Synonym diatrizoate sodium, sodium diatrizoate, sodium amidotrizoate, hypaque sodium, triombrin, triombrine, hypaque, vascoray, hypaque cysto, hpaque-cysto
PubChem CID 23672589
ChEBI CHEBI:53692
IUPAC Name sodium;3,5-diacetamido-2,4,6-triiodobenzoate
SMILES CC(=O)NC1=C(C(=C(C(=C1I)C(=O)[O-])I)NC(=O)C)I.[Na+]

Specifications

Melting Point 261.0°C to 262.0°C
Quantity 100 g
Solubility Information Solubility in water: 600g/L (20°C).
Formula Weight 635.9
Percent Purity 99%
Infrared Spectrum Authentic
Loss on Drying 7% max.
Chemical Name or Material Sodium diatrizoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.