missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dichromate Dihydrate, ACS Reagent, ≥99.5%, Honeywell Fluka™

Product Code. 15655370 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Poly Bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15655370 500 g Poly Bottle 500g
15635370 100 g Poly Bottle 100g
15645370 2.5 kg Poly Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15655370 Supplier Honeywell-Fluka Supplier No. 398063500G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

ACS Reagent,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-12-0
Molecular Formula Cr2H4Na2O9
Molecular Weight (g/mol) 297.995
MDL Number MFCD00149166
InChI Key JYDRNIYTFCBIFC-UHFFFAOYSA-N
Synonym sodium dichromate dihydrate, unii-52125xyy2a, ccris 6344, disodium chromate dihydrate, sodium dichromate vi dihydrate, sodium dichromate dihydrate vi, chromic acid h2cr2o7 , disodium salt, dihydrate, dichromic acid h2cr2o7 , disodium salt, dihydrate, dsstox_cid_12061, dsstox_rid_78903
PubChem CID 108005
IUPAC Name disodium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium;dihydrate
SMILES O.O.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[Na+].[Na+]

Specifications

Melting Point 357°C
Color Orange
Quantity 500 g
Linear Formula Na2Cr2O7 · 2H2O
UN Number UN3086
Solubility Information In water ∽2.355g/L at 20°C
Formula Weight 298g/mol
Percent Purity ≥99.5%
Odor Odorless
Packaging Poly Bottle
Appearance Crystals
Density 2.52g/cm3 at 13°C
Chemical Name or Material Sodium dichromate dihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.