missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dichromate, Dihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15508164 Shop All J.T.Baker Products
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
500g
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 15508164

Brand: Avantor J.T.Baker 367201

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 7789-12-0
Molecular Formula Cr2H4Na2O9
Molecular Weight (g/mol) 297.995
InChI Key JYDRNIYTFCBIFC-UHFFFAOYSA-N
Synonym sodium dichromate dihydrate, unii-52125xyy2a, ccris 6344, disodium chromate dihydrate, sodium dichromate vi dihydrate, sodium dichromate dihydrate vi, chromic acid h2cr2o7 , disodium salt, dihydrate, dichromic acid h2cr2o7 , disodium salt, dihydrate, dsstox_cid_12061, dsstox_rid_78903
PubChem CID 108005
IUPAC Name disodium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium;dihydrate
SMILES O.O.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[Na+].[Na+]

Specifications

Physical Form Crystals
Quantity 500 g
Formula Weight 298
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 2.35kg
Chemical Name or Material Sodium Dichromate, Dihydrate
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H330-Fatal if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H360FD-May damage fertility. May damage the unborn child.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.