missing translation for 'onlineSavingsMsg'
Learn More

Sodium dihydrogen citrate, 99%

Product Code. 11313236
Change view
Click to view available options
Quantity:
100 g
500 g
2000 g
Unit Size:
100g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11313236 100 g 100g
11323236 500 g 500g
11333236 2000 g 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11313236 Supplier Thermo Scientific Alfa Aesar Supplier No. L12557.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 18996-35-5 | C6H10NaO7 | 217.129 g/mol

Sodium dihydrogen citrate acts as a buffering agent and sequestrant. It is also employed as a reductant and stabilizer in the preparation of b>Fe3O4 platinum core/shell particles. Further, it is used in electrochemical reduction of proton at carbon fiber electrode in the presence and absence of platinum nanoparticles. In addition to this, it plays a useful role in characterization of carbon-based electrode materials as cathode in half-cell configuration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium dihydrogen citrate acts as a buffering agent and sequestrant. It is also employed as a reductant and stabilizer in the preparation of b>Fe3O4 platinum core/shell particles. Further, it is used in electrochemical reduction of proton at carbon fiber electrode in the presence and absence of platinum nanoparticles. In addition to this, it plays a useful role in characterization of carbon-based electrode materials as cathode in half-cell configuration.

Solubility
Soluble in water. Insoluble in ethanol.

Notes
Hygroscopic. Incompatible with bases, reducing agents and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18996-35-5
Molecular Formula C6H10NaO7
Molecular Weight (g/mol) 217.129
MDL Number MFCD00013067
InChI Key SLWOXBQHKZPUNY-UHFFFAOYSA-N
Synonym bicitra, pneucid, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, sodium salt, sodium dihydrogen citrate anhydrous, citric acid, sodium salt, citric acid sodium, sodium 2-hydroxy-1,2,3-propanetricarboxylate, 2-hydroxypropane-1,2,3-tricarboxylic acid; sodium, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, sodium salt 1:?
PubChem CID 131675399
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid;molecular hydrogen;sodium
SMILES [HH].C(C(=O)O)C(CC(=O)O)(C(=O)O)O.[Na]

Specifications

Melting Point 211°C to 212°C
Quantity 100 g
Beilstein 3920246
Solubility Information Soluble in water. Insoluble in ethanol.
Formula Weight 214.11
Percent Purity 99%
Chemical Name or Material Sodium dihydrogen citrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.