missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dihydrogen Orthophosphate, for Electrophoresis, Monobasic, Dihydrate, Fisher Chemical™

Product Code. 10244350 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
3 kg
Unit Size:
3kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10244350 3 kg 3kg
10457840 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10244350 Supplier Fisher Chemical Supplier No. S/P520/63
  • ONLINE EXCLUSIVE PRICE
    136.00 €
    132.00 € / 3kg
    Save 4.00 €
    2% Off
View all promotions
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

Sodium Dihydrogen Orthophosphate, for Electrophoresis, Monobasic, Dihydrate

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13472-35-0
Molecular Formula H6NaO6P
Molecular Weight (g/mol) 156.006
MDL Number 149209
InChI Key VBJGJHBYWREJQD-UHFFFAOYSA-M
Synonym sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh
PubChem CID 23673460
IUPAC Name sodium;dihydrogen phosphate;dihydrate
SMILES O.O.OP(=O)(O)[O-].[Na+]

Specifications

Melting Point 60°C
Color White
pH 4.5
Physical Form Solid
Quantity 3 kg
Formula Weight 156.01g/mol
Packaging Plastic powder jar
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.