missing translation for 'onlineSavingsMsg'
Learn More

Sodium dihydrogen phosphate dihydrate, BAKER, J.T. Baker™

Product Code. 10723181 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10723181 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10723181 Supplier JT Baker Supplier No. 1768.1000
  • ONLINE EXCLUSIVE PRICE
    61.00 €
    51.32 € / 1kg
    Save 9.68 €
    15% Off
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Laboratory Reagent

Specifications

Boiling Point ∼650°C (1013 hPa)
Melting Point 60°C
CAS 13472-35-0
Quantity 1 kg
Assay Percent Range min. 98%
Molecular Formula H6NaO6P
MDL Number MFCD00149209
Synonym Sodium phosphate monobasic dihydrate , Sodium dihydrogen orthophosphate dihydrate
InChI Key VBJGJHBYWREJQD-UHFFFAOYSA-M
SMILES O.O.[Na+].OP(O)([O-])=O
IUPAC Name sodium dihydrate dihydrogen phosphate
Molecular Weight (g/mol) 156.01
Density 1.91 g/cm³ (20°C)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.