missing translation for 'onlineSavingsMsg'
Learn More

Sodium dihydrogen phosphate monohydrate, BAKER ANALYZED™, J.T. Baker™

Product Code. 13429448 Shop All J.T.Baker Products
Change view
Click to view available options
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
13429448 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13429448 Supplier JT Baker Supplier No. 3818.1000
  • ONLINE EXCLUSIVE PRICE
    91.00 €
    86.41 € / 1kg
    Save 4.59 €
    5% Off
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Meets ACS Specifications

Specifications

CAS 10049-21-5
Quantity 1 kg
Assay Percent Range 98.0 to 102.0%
Molecular Formula H5NaO5P
MDL Number MFCD00149208
Synonym Sodium dihydrogen orthophosphate monohydrate , Sodium phosphate monobasic monohydrate
InChI Key BBMHARZCALWXSL-UHFFFAOYSA-N
SMILES O.[Na+].OP(O)(O)=O
IUPAC Name sodium phosphoric acid hydrate
Molecular Weight (g/mol) 139.00
Packaging Plastic bottle for solids
Density 0.9996 g/cm³ (20°C)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.