missing translation for 'onlineSavingsMsg'
Learn More

Sodium diphenylamine-4-sulfonate, ACS

Product Code. 11358858
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
This item is not returnable. View return policy

Product Code. 11358858

Brand: Thermo Scientific Alfa Aesar 041714.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Redox indicator | CAS: 6152-67-6 | C12H11NNaO3S | 272.27 g/mol

Redox indicatorSodium diphenylamine-4-sulfonate is used as a pH independent redox indicator used in the determination of redox potential. It is used in the preparation of poly-(aninile-co-N-(4-sulfophenyl)aniline) by polymerization with aniline. It acts as a ligand and form complexes with metal halides like lanthanum(III) chloride. It is also involved in the synthesis of gold nano crystals by reaction with chloroauric acid in the presence of polyethylene glycol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6152-67-6
Molecular Formula C12H11NNaO3S
Molecular Weight (g/mol) 272.27
MDL Number MFCD00036468
InChI Key VMVSDVIYIMNZBS-UHFFFAOYSA-N
Synonym sodium 4-phenylamino benzenesulfonate, sodium diphenylamine-4-sulfonate, benzenesulfonic acid, 4-phenylamino-, monosodium salt, unii-3e2ux1382o, sodium 4-anilinobenzenesulfonate, diphenylamine-4-sulfonic acid sodium salt, sodium diphenylamine-p-sulfonate, n-phenylsulfanilic acid, sodium salt, 4-phenylamino benzenesulfonic acid sodium salt, diphenylaminesulfonic acid sodium salt
PubChem CID 517002
IUPAC Name sodium;4-anilinobenzenesulfonate
SMILES [Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1

Specifications

Quantity 50 g
Solubility Information Soluble in water.
Formula Weight 271.27
Grade ACS
Physical Form Powder
Chemical Name or Material Sodium diphenylamine-4-sulfonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.