missing translation for 'onlineSavingsMsg'
Learn More

Sodium dodecyl sulfate, 95%, MP Biomedicals™

Product Code. 11446051 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g

Product Code. 11446051

Brand: MP Biomedicals 0219052280

Please to purchase this item. Need a web account? Register with us today!

Approximately 95% alkyl sulfate, predominantly composed of C12 and C14 homologs

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym SDS, Lauryl sulfate sodium salt
PubChem CID 3423265
ChEBI CHEBI:8984
IUPAC Name sodium dodecyl sulfate
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Color White
Physical Form Granular Powder
Density >1.1 at 20°C (water = 1)
Quantity 100 g
Assay Percent Range 0.95
Formula Weight 288.38
Chemical Name or Material Sodium dodecyl sulfate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.