missing translation for 'onlineSavingsMsg'
Learn More

Sodium dodecyl sulfate, 99.5%, MP Biomedicals™

Product Code. 17811391 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Unit Size:
100g
1kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17811391 25 g 25g
11446571 100 g 100g
11466571 500 g 500g
11476571 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17811391 Supplier MP Biomedicals Supplier No. 0219483125
  • ONLINE EXCLUSIVE PRICE
    54.25 €
    51.45 € / 25g
    Save 2.80 €
    5% Off
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym Sodium lauryl sulfate, SDS
PubChem CID 3423265
ChEBI CHEBI:8984
IUPAC Name sodium dodecyl sulfate
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Color White
Physical Form Granular Powder
Quantity 25 g
Formula Weight 288.38
Percent Purity 99.5%
Grade Molecular Biology
Chemical Name or Material Sodium dodecyl sulfate
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium Dodecyl Sulphate, Molecular Biology Reagent
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H228-Flammable solid.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Sodium Dodecyl Sulphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.