missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dodecyl Sulfate, 99+%, for Electrophoresis, Fisher Chemical™

Product Code. 10509770 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10509770 500 g 500g
10182440 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10509770 Supplier Fisher Chemical Supplier No. S/P530/53
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Sodium Dodecyl Sulfate, 99+%, for Electrophoresis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal
PubChem CID 3423265
ChEBI CHEBI:8984
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Melting Point 206°C
Color White
pH 8.5 to 10
Physical Form Solid
Quantity 500 g
IUPAC Name sodium dodecyl sulfate
Formula Weight 288.38g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dodecyl sulfate sodium salt
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H228-Flammable solid.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.