missing translation for 'onlineSavingsMsg'
Learn More

Sodium dodecyl sulfate, >99%, MP Biomedicals™

Product Code. 11446312 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
1 kg
Unit Size:
100g
10g
1kg
25g
500g

Product Code. 11446312

Brand: MP Biomedicals 0210291810

Please to purchase this item. Need a web account? Register with us today!

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal
PubChem CID 3423265
ChEBI CHEBI:8984
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Color White
Density >1.1 at 20°C
Quantity 10 g
IUPAC Name sodium dodecyl sulfate
Formula Weight 288.38
Percent Purity >99%
Physical Form Granular Powder
Chemical Name or Material Sodium dodecyl sulfate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.