missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dodecyl Sulfate (SDS), ≥95%

Product Code. 15865168 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Cardboard Carton
Plastic bottle
Unit Size:
1kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15865168 500 g Plastic bottle 500g
15865308 1 kg Plastic bottle 1kg
15875308 5 kg Cardboard Carton 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15865168 Supplier Thermo Scientific Supplier No. J18220.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym Sodium Lauryl Sulfate,n to Dodecyl sulfate sodium salt
PubChem CID 3423265
ChEBI CHEBI:8984
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Physical Form Crystalline Powder
Melting Point ∼206°C
Quantity 500 g
Packaging Plastic bottle
Linear Formula CH3(CH2)11OSO3Na
UN Number UN2926
Beilstein 3599286
IUPAC Name sodium dodecyl sulfate
Formula Weight 288.38
Percent Purity ≥95%
Chemical Name or Material Sodium Dodecyl Sulfate (SDS)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.