missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dodecyl Sulfate, Technical, Fisher Chemical

Product Code. 10090490 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10090490 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10090490 Supplier Fisher Chemical Supplier No. S/5200/53
  • ONLINE EXCLUSIVE PRICE
    122.76 €
    92.60 € / 500g
    Save 30.16 €
    24% Off
View all promotions
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

500g, Plastic powder jar

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 206°C
CAS 151-21-3
Color White
pH 8.5 to 10
Physical Form Solid
Quantity 500 g
Molecular Formula C12H25NaO4S
MDL Number MFCD00036175
Synonym sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
IUPAC Name sodium;dodecyl sulfate
Molecular Weight (g/mol) 288.38
PubChem CID 3423265
ChEBI CHEBI:8984
Formula Weight 288.38g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dodecyl sulfate sodium salt
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H228-Flammable solid.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Sodium dodecyl sulphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.