missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- For Electrophoresis and Protein Solubilization; For Laboratory,Research,or Manufacturing Use
Specifications
Specifications
| Quantity | 2 kg |
| Flash Point | 170.00°C |
| Packaging | Poly Pail |
| Specific Gravity | 1L = 0.40kg |
| IUPAC Name | sodium dodecyl sulfate |
| Formula Weight | 288.38 |
| Grade | Ultrapure Bioreagent |
| Chemical Name or Material | Sodium Dodecyl Sulfate |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 2
- Skin corrosion/irritation Category 2
- H228-Flammable solid.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P362-Take off contaminated clothing.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?