missing translation for 'onlineSavingsMsg'
Learn More

Sodium Ethoxide (>85%), TRC

Product Code. 30410119
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30410119 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30410119 Supplier Toronto Research Chemicals Supplier No. TRCS62500010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Ethoxide
CAS 141-52-6
Molecular Formula C2H5NaO
Quantity 10 g
Synonym Ethanol Sodium Salt, Ethyl Alcohol Sodium Salt, Caustic Alcohol, Ethoxysodium, Sodium Ethanolate, Sodium Ethoxylate, Sodium Ethylate
InChI Formula InChI=1S/C2H5O.Na/c1-2-3;/h2H2,1H3;/q-1;+1
SMILES [Na+].CC[O-]
IUPAC Name sodium;ethanolate
Molecular Weight (g/mol) 68.05
Formula Weight 68.0238
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.