missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Ethyl Sulfate, TRC
Specifications
Specifications
| Chemical Name or Material | Sodium Ethyl Sulfate |
| CAS | 546-74-7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C2H5NaO4S |
| Quantity | 500 mg |
| Synonym | Sulfuric Acid Monoethyl Ester Sodium Salt, Ethyl Sodium Sulfate, Sodium Sulfovinate |
| InChI Formula | InChI=1S/C2H6O4S.Na/c1-2-6-7(3,4)5;/h2H2,1H3,(H,3,4,5);/q;+1/p-1 |
| SMILES | CCOS(=O)(=O)[O-].[Na+] |
| IUPAC Name | sodium;ethyl sulfate |
| Molecular Weight (g/mol) | 148.11 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?