missing translation for 'onlineSavingsMsg'
Learn More

Sodium ferrocyanide decahydrate

Product Code. 15560614 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
Product Code. Quantity unitSize
15560614 5 kg 5kg
15570614 250 g 250g
15550614 1 kg 1kg
3 options
This item is not returnable. View return policy

Product Code. 15560614

Brand: Thermo Scientific Acros 459290050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14434-22-1 | C6H20FeN6Na4O10 | 484.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14434-22-1
Molecular Formula C6H20FeN6Na4O10
Molecular Weight (g/mol) 484.06
MDL Number MFCD00149274
InChI Key JFSUDVTVQZUDOP-UHFFFAOYSA-N
SMILES O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].[Fe++].[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N

Specifications

Quantity 5 kg
Solubility Information Solubility in water: 369g/L (20°C). Other solubilities: insoluble in alcohol
Formula Weight 484.06
Loss on Drying 1% max. (free moisture)
Chemical Name or Material Sodium ferrocyanide decahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.