missing translation for 'onlineSavingsMsg'
Learn More

Sodium fluoride, 200mM aq. soln., Thermo Scientific Chemicals

Product Code. 15463049
Change view
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
Product Code. Quantity unitSize
15463049 250 mL 250mL
15453049 100 mL 100mL
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15463049

Brand: Thermo Scientific Alfa Aesar J60251.AK

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-49-4
Molecular Formula FNa
Molecular Weight (g/mol) 41.99
MDL Number MFCD00003524
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
Synonym sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril
PubChem CID 5235
ChEBI CHEBI:28741
IUPAC Name sodium fluoride
SMILES [F-].[Na+]

Specifications

Color Colorless
Physical Form Liquid
Quantity 250 mL
Linear Formula NaF
Formula Weight 41.99
Concentration or Composition (by Analyte or Components) 200mM aq. soln.
Odor Odorless
Chemical Name or Material Sodium fluoride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.