missing translation for 'onlineSavingsMsg'
Learn More

Sodium fluoride, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 15402807
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15402807 10 g 10g
15412807 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15402807 Supplier Thermo Scientific Alfa Aesar Supplier No. 046912.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Regulatorischer Hinweis: Abgabe sachkundepflichtiger Stoffe und Gemische nur bei Erfüllung der Vorrausetzungen nach Chemikalienverbotsverordnung (ChemVerbotsV). Keine Abgabe an Privatpersonen.

Sodium fluoride is used in laundry souring, manufacturing enamels, manufacturing coated papers, wood preservative , foam production, opal glass, ore flotation and water fluoridation. It is also acts as an inhibitor of serine/threonine phosphatases widely used in bone and dental research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 7681-49-4
Molecular Formula FNa
Molecular Weight (g/mol) 41.99
MDL Number MFCD00003524
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
Synonym sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril
PubChem CID 5235
ChEBI CHEBI:28741
IUPAC Name sodium fluoride
SMILES [F-].[Na+]

Spezifikation

Boiling Point 1704°C
Melting Point 993°C
Color White
Physical Form Pieces
Quantity 10 g
Linear Formula NaF
UN Number UN1690
Merck Index 14,8618
Solubility Information It is soluble in water,slightly soluble in HF,ammonia. Negligible in alcohol,acetone,SO2,dimethylformamide.
Formula Weight 41.99
Percent Purity 99.99%
Odor Odorless
Refractive Index 1.336
Density 2.78 g/mL
Assay (metals basis)
Chemical Name or Material Sodium fluoride
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • Sodium fluoride
Signalord
  • Danger
Farekategori
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Faresætning
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • EUH032-Contact with acids liberates very toxic gas.
Sikkerhedssætning
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplerende oplysninger
  • MIXTURE LIST-Contain: Sodium fluoride

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.