missing translation for 'onlineSavingsMsg'
Learn More

Sodium fluoride, ≥99%, ACS reagent, powder, Thermo Scientific Chemicals

Product Code. 10582764 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
2.5kg
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10582764 2.5 kg Plastic bottle 2.5kg
10742222 5 g Plastic bottle 5g
10405291 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10582764 Supplier Thermo Scientific Acros Supplier No. 424320025
  • ONLINE EXCLUSIVE PRICE
    265.00 €
    248.00 € / 2.5kg
    Save 17.00 €
    6% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-49-4
Molecular Formula FNa
Molecular Weight (g/mol) 41.99
MDL Number MFCD00003524
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
Synonym floridine, florocid, sodium fluoride, sodium fluoride naf, zymafluor, fluonatril, ossin, antibulit, sodium fluoride, fluoride, sodium
PubChem CID 5235
ChEBI CHEBI:28741
SMILES [F-].[Na+]

Specifications

Boiling Point 1700.0°C
Melting Point 993.0°C
Color White
Physical Form Powder
Quantity 2.5 kg
Assay Percent Range ≥99%
Linear Formula NaF
Fieser 01,1073; 02,382
Merck Index 15, 8754
Solubility Information Solubility aq. soln.: 40 g/L (25°C). Other solubilities: slightly soluble in alcohol
IUPAC Name sodium fluoride
Formula Weight 41.98
Percent Purity ≥99%
Grade ACS Reagent
Loss on Drying 0.3% max. (150°C)
Packaging Plastic bottle
Chemical Name or Material Sodium fluoride
Show More Show Less
compliance-icons
Product Identifier
  • Sodium fluoride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.