missing translation for 'onlineSavingsMsg'
Learn More

Sodium formaldehydesulfoxylate dihydrate, 98%

Product Code. 10746351 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
Unit Size:
1kg
25kg
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10746351 1 kg Plastic bottle 1kg
10589053 5 kg Plastic bottle 5kg
11994221 25 kg Fibre drum 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10746351 Supplier Thermo Scientific Acros Supplier No. 149190010
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 6035-47-8 | CH3NaO32H2O | 154.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6035-47-8
Molecular Formula CH3NaO32H2O
Molecular Weight (g/mol) 154.11
MDL Number MFCD00150598
InChI Key UCWBKJOCRGQBNW-UHFFFAOYSA-M
Synonym sodium hydroxymethanesulfinate dihydrate, sodium formaldehydesulfoxylate dihydrate, methanesulfinic acid, hydroxy-, monosodium salt, dihydrate, sodium formaldehyde sulfoxylate, sodium dihydrate hydroxymethanesulfinate, sodium formaldehyde sulfoxylate nf, formaldehyde sodium sulfoxylate dihydrate, monosodium hydroxymethanesulfinate dihydrate, 79-25-4 parent, hydroxymethanesulfinic acid monosodium salt dihydrate
PubChem CID 23666330
IUPAC Name sodium;hydroxymethanesulfinate;dihydrate
SMILES C(O)S(=O)[O-].O.O.[Na+]

Specifications

Melting Point 63.0°C to 64.0°C
Assay Percent Range 97.5% min. (Iodometry)
Packaging Plastic bottle
Linear Formula HOCH2SO2Na·2H2O
Quantity 1 kg
Merck Index 15,8755
Solubility Information Solubility in water: 1088g/L. Other solubilities: practically insoluble in abs.alcohol, ether and, benzene
Formula Weight 154.11
Percent Purity 98%
Chemical Name or Material Sodium formaldehydesulfoxylate dihydrate
compliance-icons
Product Identifier
  • Sodium formaldehydesulfoxylate dihydrate
Signal Word
  • Warning
Hazard Category
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H361d-Suspected of damaging the unborn child.
  • H341-Suspected of causing genetic defects.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.