missing translation for 'onlineSavingsMsg'
Learn More

Sodium Formate ACS MP Biomedicals

Product Code. 11427882 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11427882 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11427882 Supplier MP Biomedicals Supplier No. 0215257690
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 141-53-7
Quantity 500 g
Molecular Formula CHNaO2
Synonym sodium formate, formic acid, sodium salt, salachlor, formic acid sodium salt, formic acid, na salt, sodium formate, refined, sodium formiate, mravencan sodny czech, ccris 1037, hsdb 744
InChI Key HLBBKKJFGFRGMU-UHFFFAOYSA-M
SMILES C(=O)[O-].[Na+]
IUPAC Name sodium;formate
Molecular Weight (g/mol) 68.007
PubChem CID 2723810
ChEBI CHEBI:62965
Formula Weight 68.02
Grade ACS
Density 1.92 g/cm3 at 20 °C
Chemical Name or Material Sodium Formate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.