missing translation for 'onlineSavingsMsg'
Learn More

Sodium Formate, purum p.a, ≥98.0% (NT), Honeywell™

Product Code. p-7120791 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Poly bottle
Unit Size:
1kg
250g
5kg
This item is not returnable. View return policy

Product Code. 15626800

Brand: Honeywell Research Chemicals 71540250G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum p.a.,≥98.0% (NT)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13755-29-8
Molecular Formula BF4Na
Molecular Weight (g/mol) 109.79
MDL Number MFCD00003515
InChI Key KGJZTOFHXCFQIV-UHFFFAOYSA-N
Synonym sodium tetrafluoroborate, sodium fluoborate, sodium fluoroborate, sodium borofluoride, sodium boron fluoride, apreton r, sodium boron tetrafluoride, sodium fluoroborate nabf4, sodium tetrafluoroborate 1-, sodium tetrafluroborate
PubChem CID 4343483
SMILES [Na+].F[B-](F)(F)F

Specifications

Melting Point 255°C
Color Colorless
pH 9.0 to 10.0 (at 20°C)
Physical Form Solid
Quantity 250 g
Linear Formula NaBF4
UN Number NONH for all modes of transport
Solubility Information 700g/L (at 20°C)
IUPAC Name sodium tetrafluoroboranuide
Formula Weight 109.79g/mol
Percent Purity ≥98.0% (T)
Packaging Poly bottle
Density 2.47g/mL at 25°C
Chemical Name or Material Sodium tetrafluoroborate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.