missing translation for 'onlineSavingsMsg'
Learn More

Sodium Formate, Reagent grade, 97%, Honeywell™

Product Code. p-7120792 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
4 kg
Packaging:
Poly Bottle
Unit Size:
1kg
4kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15640980 1 kg Poly Bottle 1kg
15650980 4 kg Poly Bottle 4kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15640980 Supplier Honeywell Research Chemicals Supplier No. 1076031KG
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, 97%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-53-7
Molecular Formula CHNaO2
Molecular Weight (g/mol) 68.007
MDL Number MFCD00013101
InChI Key HLBBKKJFGFRGMU-UHFFFAOYSA-M
Synonym sodium formate, formic acid, sodium salt, salachlor, formic acid sodium salt, formic acid, na salt, sodium formate, refined, sodium formiate, mravencan sodny czech, ccris 1037, hsdb 744
PubChem CID 2723810
ChEBI CHEBI:62965
IUPAC Name sodium;formate
SMILES C(=O)[O-].[Na+]

Specifications

Melting Point 255°C
Color Colorless
pH 9 to 10 (at 20°C)
Physical Form Solid
Quantity 1 kg
Linear Formula HCOONa
UN Number NONH for all modes of transport
Beilstein 3595134
Solubility Information Soluble in water
Formula Weight 68.01g/mol
Percent Purity 97%
Grade Reagent
Packaging Poly Bottle
Density 1.92g/cu.cm
Chemical Name or Material Sodium Formate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.