missing translation for 'onlineSavingsMsg'
Learn More

Sodium Formate, TRC

Product Code. 30411488
Change view
Click to view available options
Quantity:
5 g
500 g
Unit Size:
500g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30411488 500 g 500g
30426686 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30411488 Supplier Toronto Research Chemicals Supplier No. TRCS629000500G
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-53-7
Molecular Formula CHNaO2
Molecular Weight (g/mol) 68.01
Synonym Formic Acid Sodium Salt, NA-Amasil
IUPAC Name sodium;formate
SMILES [Na+].[O-]C=O

Specifications

Quantity 500 g
InChI Formula InChI=1S/CH2O2.Na/c2-1-3;/h1H,(H,2,3);/q;+1/p-1
Formula Weight 67.9874
Chemical Name or Material Sodium Formate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.