missing translation for 'onlineSavingsMsg'
Learn More

Sodium fumarate, 98%

Product Code. 11403634
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403634 100 g 100g
11413634 500 g 500g
11423634 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11403634 Supplier Thermo Scientific Alfa Aesar Supplier No. A11276.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17013-01-3 | C4H4Na2O4 | 162.052 g/mol

Sodium fumarate is used as an acidity regulator in food processing and as terminal electron acceptors in the cultivation of certain anaerobic microorganisms. It is also used as a buffering agent and pH adjuster in cosmetic and personal care products. Further, it is used as a food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium fumarate is used as an acidity regulator in food processing and as terminal electron acceptors in the cultivation of certain anaerobic microorganisms. It is also used as a buffering agent and pH adjuster in cosmetic and personal care products. Further, it is used as a food additive.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17013-01-3
Molecular Formula C4H4Na2O4
Molecular Weight (g/mol) 162.052
MDL Number MFCD00064567
InChI Key VXXVUHAXJHEYFH-SEPHDYHBSA-N
Synonym disodium fumarate, sodium fumarate, sodium fumarate dibasic, fumaric acid, disodium salt, fumaric acid disodium salt, fumaran sodny czech, fumaran sodny, disodium butenedioate, di-sodium fumarate, sodium monofumarate
PubChem CID 87110070
IUPAC Name (E)-but-2-enedioic acid;sodium
SMILES C(=CC(=O)O)C(=O)O.[Na].[Na]

Specifications

Assay Percent Range 98%
Quantity 100 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 160.04
Percent Purity 98%
Chemical Name or Material Sodium fumarate
compliance-icons
Product Identifier
  • Sodium fumarate
Signal Word
  • Warning
Catégorie de danger
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Conseils de prudence
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Informations supplémentaires
  • MIXTURE LIST-Contain: Sodium fumarate

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.