missing translation for 'onlineSavingsMsg'
Learn More

Sodium fumarate, 98%

Product Code. 11413634
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11413634 500 g 500g
11403634 100 g 100g
11423634 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11413634 Supplier Thermo Scientific Alfa Aesar Supplier No. A11276.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 17013-01-3 | C4H4Na2O4 | 162.052 g/mol

Sodium fumarate is used as an acidity regulator in food processing and as terminal electron acceptors in the cultivation of certain anaerobic microorganisms. It is also used as a buffering agent and pH adjuster in cosmetic and personal care products. Further, it is used as a food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium fumarate is used as an acidity regulator in food processing and as terminal electron acceptors in the cultivation of certain anaerobic microorganisms. It is also used as a buffering agent and pH adjuster in cosmetic and personal care products. Further, it is used as a food additive.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17013-01-3
Molecular Formula C4H4Na2O4
Molecular Weight (g/mol) 162.052
MDL Number MFCD00064567
InChI Key VXXVUHAXJHEYFH-SEPHDYHBSA-N
Synonym disodium fumarate, sodium fumarate, sodium fumarate dibasic, fumaric acid, disodium salt, fumaric acid disodium salt, fumaran sodny czech, fumaran sodny, disodium butenedioate, di-sodium fumarate, sodium monofumarate
PubChem CID 87110070
IUPAC Name (E)-but-2-enedioic acid;sodium
SMILES C(=CC(=O)O)C(=O)O.[Na].[Na]

Specifications

Assay Percent Range 98%
Quantity 500 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 160.04
Percent Purity 98%
Chemical Name or Material Sodium fumarate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.