missing translation for 'onlineSavingsMsg'
Learn More

Sodium galacturonate anhydrous, 98%

Product Code. 15408189
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15408189

Brand: Thermo Scientific Alfa Aesar J66724.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14984-39-5 | C6H9NaO7 | 216.121 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14984-39-5
Molecular Formula C6H9NaO7
Molecular Weight (g/mol) 216.121
MDL Number MFCD06201013
InChI Key MSXHSNHNTORCAW-NSBQNPTMSA-M
Synonym d-+-galacturonic acid sodium salt
PubChem CID 122174033
IUPAC Name sodium;(4S)-3,4,5,6-tetrahydroxyoxane-2-carboxylate
SMILES C1(C(C(OC(C1O)O)C(=O)[O-])O)O.[Na+]

Specifications

Color White
Physical Form Powder
Quantity 5 g
Formula Weight 216.12
Percent Purity 98%
Sensitivity Hygroscopic
Chemical Name or Material Sodium galacturonate, anhydrous

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.