missing translation for 'onlineSavingsMsg'
Learn More

Sodium glucoheptonate

Product Code. 15458319
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15458319 500 g 500g
15448319 100 g 100g
15468319 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15458319 Supplier Thermo Scientific Alfa Aesar Supplier No. J66811.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 31138-65-5 | C7H13NaO8 | 248.163 g/mol

Sodium glucoheptonate is used for cleaning metal, mercerizing, paint stripping, and aluminum etching. It is also used to mercerize cotton, linen, hemp, and other plant-based fibers to increase strength and add luster to their overall appearance of threads and fabrics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium glucoheptonate is used for cleaning metal, mercerizing, paint stripping, and aluminum etching. It is also used to mercerize cotton, linen, hemp, and other plant-based fibers to increase strength and add luster to their overall appearance of threads and fabrics.

Solubility
Freely soluble in water

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31138-65-5
Molecular Formula C7H13NaO8
Molecular Weight (g/mol) 248.163
MDL Number MFCD00013425
InChI Key FMYOMWCQJXWGEN-AXUKCSBQSA-M
Synonym sodium glucoheptonate, sodium 2r,3r,4r,5s,6r-2,3,4,5,6,7-hexahydroxyheptanoate, d-sodium glucoheptonate, sodium-alpha-glucoheptonate, 2r,3r,4r,5s,6r-2,3,4,5,6,7-hexahydroxyheptanoic acid sodium salt
PubChem CID 49853507
IUPAC Name sodium;(2R,3R,4R,5S,6R)-2,3,4,5,6,7-hexahydroxyheptanoate
SMILES C(C(C(C(C(C(C(=O)[O-])O)O)O)O)O)O.[Na+]

Specifications

Color White
Physical Form Powder
Quantity 500 g
Solubility Information Freely soluble in water
Formula Weight 248.16
Chemical Name or Material Sodium glucoheptonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.